Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MSP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144959
Description
Immunogen sequence: PEWYVVPPGT KCEIAGWGET KGTGNDTVLN VALLNVISNQ ECNIKHRG Antigen sequence identity: Mouse 79%; Rat 73%.
MSP (or MST1) contains four kringle domains and a serine protease domain, similar to that found in hepatic growth factor. Despite the presence of the serine protease domain, the encoded protein may not have any proteolytic activity. The receptor for this protein is RON tyrosine kinase, which upon activation stimulates ciliary motility of ciliated epithelial lung cells. This protein is secreted and cleaved to form an alpha chain and a beta chain bridged by disulfide bonds.
Specifications
MSP | |
Polyclonal | |
Unconjugated | |
MST1 | |
D3F15S2; D3F15S2h; D8h3f15s2; D9H3F15S2; DNF15S2; DNF15S2h; E2F transcription factor 2; E2F2; Hepatocyte growth factor-like protein; Hepatocyte growth factor-like protein alpha chain; Hepatocyte growth factor-like protein beta chain; hepatocyte growth factor-like protein homolog; Hgfl; macrophage stimulating 1; macrophage stimulating 1 (hepatocyte growth factor-like); macrophage stimulatory protein; Macrophage-stimulating protein; MSP; Mst1; NF15S2; Unknown (protein for MGC:137619) | |
Rabbit | |
Affinity chromatography | |
RUO | |
4485 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P26927 | |
MST1 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 591-638. Recombinant protein control fragment (Product #RP-110126). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction