Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588894 Supplier Invitrogen™ Supplier No. PA588894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CREAACVWCN GEEYRGAVDR TESGRECQRW DLQHPHQHPF EPGKFLDQGL DDNYCRNPDG SERPWCYTTD PQIEREFCDL PRCGSEAQPR QEATTVSCFR GKGEGYRGTA NTTTAGVPCQ RWDAQIPHQH RFTPEKYACK DLRENFCRNP DGSEAPWCFT LRPGMRAAFC YQIRRCTDDV RPQDCYHGAG EQYRGTVSKT RKGVQCQRWS AETPHKPQFT FTSEPHAQLE ENFCRNPDGD SHGPWCYTMD PRTPFDYCAL RRCADDQPPS ILDPPDQVQF EKCGKRVDRL DQRRSKLRVV G; Positive Samples: Mouse kidney, Rat kidney.

MSP (or MST1) contains four kringle domains and a serine protease domain, similar to that found in hepatic growth factor. Despite the presence of the serine protease domain, the encoded protein may not have any proteolytic activity. The receptor for this protein is RON tyrosine kinase, which upon activation stimulates ciliary motility of ciliated epithelial lung cells. This protein is secreted and cleaved to form an alpha chain and a beta chain bridged by disulfide bonds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MST1
Gene Accession No. P26927, P26928
Gene Alias D3F15S2; D3F15S2h; D8h3f15s2; D9H3F15S2; DNF15S2; DNF15S2h; E2F transcription factor 2; E2F2; Hepatocyte growth factor-like protein; Hepatocyte growth factor-like protein alpha chain; Hepatocyte growth factor-like protein beta chain; hepatocyte growth factor-like protein homolog; Hgfl; macrophage stimulating 1; macrophage stimulating 1 (hepatocyte growth factor-like); macrophage stimulatory protein; Macrophage-stimulating protein; MSP; Mst1; NF15S2; Unknown (protein for MGC:137619)
Gene Symbols MST1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-500 of human MSP/MST1 (NP_0662783).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15235, 24566, 4485
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.