Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSR1 (CD204) Mouse anti-Human, Brilliant Violet 786, Clone: U23-56, BD Optibuild™

Catalog No. BDB742443 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742443 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742443 Supplier BD Biosciences Supplier No. 742443
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The U23-56 monoclonal antibody specifically binds to Macrophage scavenger receptor 1 (MSR1) which is also known as CD204, Scavenger receptor A (SRA), Scavenger receptor class A member 1 (SCARA1), or Macrophage acetylated LDL receptor I and II. MSR1 is a trimeric type II transmembrane glycoprotein that belongs to the type 1 class A scavenger receptor family. MSR1 functions as a scavenger receptor that enables mononuclear phagocytes to take up and respond to a variety of negatively charged macromolecules including oxidized and acetylated low density lipoproteins. MSR1 also functions in innate immunity as a pattern recognition receptor. MSR1 allows macrophages to recognize and respond to microbial products, such as lipopolysaccharide and polyribonucleic acids, by the production of cytokines and inflammatory mediators. MSR1 also reportedly serves as a coreceptor that regulates the signaling of some Toll-like receptors including TLR-3, TLR-4, and TLR-9. MSR1 has been associated with several diseases including atherosclerosis, Alzheimer’s disease, ischemic injury, and sepsis. The U23-56 antibody was tested by flow cytometric and Western blot analyses of MSR1-transfected and untransfected cells to verify its specificity.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSR1 (CD204)
Classification Monoclonal
Clone U23-56
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias MSR, MSRE, phSR1, phSR2, SCARA1, SR-A, SRA
Host Species Mouse
Immunogen Human MSR1 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.