Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSRA Mouse anti-Human, Clone: 3C11, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation H00004482M01A
Description
Mouse monoclonal antibody raised against a partial recombinant MSRA.
- High specificity
- High reactivity
Specifications
MSRA | |
Cocktail | |
Unconjugated | |
ascites with no preservative | |
NM_012331 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
Ig Mix |
ELISA, Western Blot | |
3C11 | |
Mouse monoclonal antibody raised against a partial recombinant MSRA. | |
MSRA | |
MSRA | |
MSRA (NP_036463.1, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
4482 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction