Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSRA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA514206
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA514206 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA514206 Supplier Invitrogen™ Supplier No. PA514206
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine based on sequence homology.

Methionine sulfoxide reductase (MsrA) reduces methionine sulfoxide (MetO) residues in proteins and free MetO to Methionine (Met). The catalytic activity of MsrA is dependent of bound metal and cofactors but it requires reducing equivalents from either DTT or a thioredoxin-regenerating system. MsrA plays an essential role in protecting cells against oxidative damage. The substrates of MsrA include calmodulin, HIV protease and a 1-proteinase-inhibitor. Recent studies indicate that there is a connection between MsrA and Alzheimer's disease in mammals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSRA
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Msra
Gene Accession No. Q9D6Y7, Q9UJ68
Gene Alias 2310045J23Rik; 6530413P12Rik; cytosolic methionine-S-sulfoxide reductase; methionine sulfoxide reductase A; mitochondrial peptide methionine sulfoxide reductase; Msra; MSR-A; peptide met (O) reductase; peptide Met(O) reductase; peptide methionine sulfoxide reductase; peptide-methionine (S)-S-oxide reductase; PMSR; protein-methionine-S-oxide reductase
Gene Symbols Msra
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 34-63 amino acids from the N-terminal region of human MSRA.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110265, 4482
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.