Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSRB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565360
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565360 Supplier Invitrogen™ Supplier No. PA565360
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVQMWETGQVS CTGSRTKVNL LCRRSAWEMG VWEETPSASP SKGLHCGCIM ARSSPSCLVI FPGVLWQVWQ WVGPRVPERR PQAGAVPILN IQQLAEVCP Highest antigen sequence identity to the following orthologs - mouse 27%, rat 24%.

This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. This protein belongs to the methionine sulfoxide reductase B (MsrB) family, and it is expressed in a variety of adult and fetal tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSRB1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MSRB1
Gene Accession No. Q9NZV6
Gene Alias D17Wsu82e; HSPC270; methionine sulfoxide red. domain; methionine sulfoxide reductase b; methionine sulfoxide reductase B1; methionine-R-sulfoxide reductase B1; MGC3344; MsrB1; Selenoprotein R; Selenoprotein X; selenoprotein X 1; selenoprotein X, 1; SelR; SELX; Sepr; Sepx1
Gene Symbols MSRB1
Host Species Rabbit
Immunogen Recombinant Human MSRB1. Recombinant protein control fragment (Product #RP-106042).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51734
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.