Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSRB1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

$209.50 - $511.50

Specifications

Antigen MSRB1
Concentration 0.5 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5121763
View Documents
Invitrogen™
PA5121763
20 μL
Each for $209.50
Only null left
Add to Cart
 
PIPA5121762
View Documents
Invitrogen™
PA5121762
100 μL
Each for $511.50
Only null left
Add to Cart
 
Description

Description

This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. This protein belongs to the methionine sulfoxide reductase B (MsrB) family, and it is expressed in a variety of adult and fetal tissues.
Specifications

Specifications

MSRB1
Western Blot
Unconjugated
Rabbit
Human
Q9NZV6
51734
Met1-His116 (Accession Q9NZV6), with N-terminal His Tag.
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
MSRB1
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
D17Wsu82e; HSPC270; methionine sulfoxide red. domain; methionine sulfoxide reductase b; methionine sulfoxide reductase B1; methionine-R-sulfoxide reductase B1; MGC3344; MsrB1; Selenoprotein R; Selenoprotein X; selenoprotein X 1; selenoprotein X, 1; SelR; SELX; Sepr; Sepx1
MSRB1
IgG
Antigen affinity chromatography, Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.