Learn More
Invitrogen™ MSRB2 Monoclonal Antibody (3F12)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA521370
Catalog No. PIMA521370
Only null left
Add to Cart
Description
Description
Peptide Sequence: KEAGMYHCVC CDSPLFSSEK KYCSGTGWPS FSEAHGTSGS DESHTGILRR LDTSLGSART EVVCKQCEAH LGHVFPDGPG PNGQRFCINS VALKFKPRKH.
MSRB2 catalyzes the reduction of free and protein-bound methionine sulfoxide to methionine.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MSRB2
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9Y3D2
MSRB2
MSRB2 (NP_036360, 102 a.a. ~ 201 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG1 κ
ELISA, Western Blot
3F12
Unconjugated
MSRB2
2310050L06Rik; CBS1; CBS-1; CGI-131; methionine sulfoxide reductase B2; methionine-R-sulfoxide reductase B2, mitochondrial; Mrsb; Msrb; MsrB2; PILB; pilin-like transcription factor
Mouse
Affinity chromatography
RUO
22921
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.