Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSRB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117128
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117128 Supplier Invitrogen™ Supplier No. PA5117128
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse heart, Mouse kidney, Mouse brain, Rat liver Immunogen sequence: GLTLGTAPRR AVRGQAGGGG PGTGPGLGEA GSLATCELPL AKSEWQKKLT PEQFYVTREK GTEPPFSGIY LNNKEAGMYH CVCCDSPLFS SEKKYCSGTG WPSFSEAHGT SGSDESHTGI LRRLDTSLGS ARTEVVCKQC EAHLGHVFPD GPGPNGQRFC INSVALKFKP RKH.

MSRB2 catalyzes the reduction of free and protein-bound methionine sulfoxide to methionine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSRB2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MSRB2
Gene Accession No. Q4FZX5, Q78J03, Q9Y3D2
Gene Alias 2310050L06Rik; CBS1; CBS-1; CGI-131; methionine sulfoxide reductase B2; methionine-R-sulfoxide reductase B2, mitochondrial; Mrsb; Msrb; MsrB2; PILB; pilin-like transcription factor
Gene Symbols MSRB2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 10-182 of human MSRB2 (NP_0363603).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22921, 361286, 76467
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.