Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MST1/2 Rabbit anti-Human, Clone: SC05-83, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267624
Description
MST1/2 Monoclonal specifically detects MST1/2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
MST1/2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin | |
D3F15S2, DNF15S2, HGFL, MSP, MST1 macrophage stimulating 1 (hepatocyte growth factor-like), NF15S2 | |
Rabbit | |
Protein A purified | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
SC05-83 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
STK3 | |
Synthetic peptide within N-terminal human MST1/2. (SwissProt: Q13188 Human; SwissProt: Q9JI11 Mouse; SwissProt: O54748 Rat) | |
100 μL | |
6788 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction