Learn More
Promega MST1 Kinase Enzyme System, 10microg
Catalog No. PRV4152 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4152 10 μg
1 options
Catalog No. PRV4152 Supplier Promega Supplier No. V4152
Recombinant full-length human MST1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MST1 belongs to a family of proteins that share similarity with a budding yeast serine/threonine kinase, sterile-20 (Ste20).
  • Easily Screen and Profile MST1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C