Learn More
Invitrogen™ MSP Monoclonal Antibody (OTI1A10)

Description
MSP Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | MSP |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1A10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MST1 |
| Gene Accession No. | P26927, P26928 |
| Gene Alias | D3F15S2; D3F15S2h; D8h3f15s2; D9H3F15S2; DNF15S2; DNF15S2h; E2F transcription factor 2; E2F2; Hepatocyte growth factor-like protein; Hepatocyte growth factor-like protein alpha chain; Hepatocyte growth factor-like protein beta chain; hepatocyte growth factor-like protein homolog; Hgfl; macrophage stimulating 1; macrophage stimulating 1 (hepatocyte growth factor-like); macrophage stimulatory protein; Macrophage-stimulating protein; MSP; Mst1; NF15S2; Unknown (protein for MGC:137619) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.