Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MST1/STK4 Antibody (01), Alexa Fluor™ 594 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306376AF594
Description
MST1/STK4 Monoclonal antibody specifically detects MST1/STK4 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
MST1/STK4 | |
Monoclonal | |
Alexa Fluor 594 | |
Mouse | |
Protein A purified | |
RUO | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence | |
01 | |
50mM Sodium Borate | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MST1/STK4 (Q13043-1; Glu2-Phe487). | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction