Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MST1/STK4 Antibody (01), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306376100UL
Description
MST1/STK4 Monoclonal antibody specifically detects MST1/STK4 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ ImmunofluorescenceSpecifications
MST1/STK4 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Flow Cytometry, Immunofluorescence | |
01 | |
Flow Cytometry 1:25-1:100, Immunocytochemistry/ Immunofluorescence 1:20-1:100 | |
AI447339;AU020804;Kas-2;KRS2;MST1;STK4;YSK3 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MST1/STK4 (Uniprot#: Q13043-1; Glu2-Phe487). | |
100 μg | |
Apoptosis, Autophagy, Cardiovascular Biology, Cell Biology, Signal Transduction | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction