Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MST1/STK4 Antibody (07), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306389MFV500
Description
MST1/STK4 Monoclonal antibody specifically detects MST1/STK4 in Human samples. It is validated for ELISA, ELISASpecifications
MST1/STK4 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Human | |
Purified |
ELISA, Sandwich ELISA | |
07 | |
ELISA, Sandwich ELISA Capture | |
AI447339;AU020804;Kas-2;KRS2;MST1;STK4;YSK3 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MST1/STK4 (Uniprot#: Q13043-1; Glu2-Phe487). | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction