Learn More
Invitrogen™ Human MST1R (RON), GST Tag Recombinant Protein

Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic

Specifications
Specifications
Accession Number | Q04912 |
Concentration | See Label |
For Use With (Application) | Kinase Assay |
Formulation | 50 mM tris HCl with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
Gene ID (Entrez) | 4486 |
Molecular Weight (g/mol) | 74.8 kDa |
Name | Human MST1R (RON), GST Tag |
Quantity | 10 μg |
Regulatory Status | For research use only. Not for use in diagnostic procedures |
Gene Alias | CD136; Cdw136; c-met-related tyrosine kinase; friend virus susceptibility 2; Fv2; Fv-2; macrophage stimulating 1 receptor; macrophage stimulating 1 receptor (c-met-related tyrosine kinase); macrophage-stimulating protein receptor; Macrophage-stimulating protein receptor alpha chain; Macrophage-stimulating protein receptor beta chain; MSP receptor; Mst1r; MST1R variant RON30; MST1R variant RON62; p185-Ron; protein tyrosine kinase 8; Protein-tyrosine kinase 8; PTK8; PTK8 protein tyrosine kinase 8; receptor protein tyrosine kinase, c-met-related; RON; RON variant E2E3; Stem cell-derived tyrosine kinase; STK |
Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.