Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MST2 Rabbit anti-Human, Zebrafish, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572649
Description
Stress-activated, pro-apoptotic kinase which, following caspase-cleavage, enters the nucleus and induces chromatin condensation followed by internucleosomal DNA fragmentation. Key component of the Hippo signaling pathway which plays a pivotal role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein stk3/mst2 and stk4/mst1, in complex with its regulatory protein sav1, phosphorylates and activates lats1/2 in complex with its regulatory protein mob1, which in turn phosphorylates and inactivates yap1 oncoprotein and wwtr1/taz. Phosphorylation of yap1 by lats2 inhibits its translocation into the nucleus to regulate cellular genes important for cell proliferation, cell death, and cell migration (By similarity).
Specifications
MST2 | |
Polyclonal | |
Unconjugated | |
STK3 | |
Serine/threonine-protein kinase 3; Serine/threonine-protein kinase 3 36kDa subunit; MST2/N; Serine/threonine-protein kinase 3 20kDa subunit; MST2/C; stk3 | |
Rabbit | |
Protein A | |
RUO | |
324125, 6788 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q13188, Q7ZUQ3 | |
STK3 | |
KLH conjugated synthetic peptide between 228-252 amino acids from the Central region of zebrafish MST2 | |
400 μL | |
Primary | |
Human, Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction