Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MST3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$416.50 - $682.00
Specifications
Antigen | MST3 |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml, ELISA, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MST3 Polyclonal specifically detects MST3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MST3 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
EC 2.7.11, EC 2.7.11.1, Mammalian STE20-like protein kinase 3, MST-3, MST3serine/threonine kinase 24 (Ste20, yeast homolog), serine/threonine kinase 24, serine/threonine-protein kinase 24, STE20, STE20-like kinase 3, STE20-like kinase MST3, sterile 20-like kinase 3, STK3yeast) | |
STK24 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.04-0.4 ug/ml, ELISA, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Protein Kinase | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
8428 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:IDRYKRWKAEQSHDDSSSEDSDAETDGQASGGSDSGDWIFTIREKDPKNLENGALQPSDLDRNKMKDIPKRPFS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title