Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MST3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517285
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517285 Supplier Invitrogen™ Supplier No. PA517285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

STK24 (MST3) is a serine/threonine kinase that is a member of the GCK subfamily of STE20-like proteins. The yeast 'Sterile 20' gene (STE20) functions upstream of the mitogen-activated protein kinase (MAPK) cascade. In mammals, protein kinases related to STE20 can be divided into 2 subfamilies based on their structure and regulation. Members of the PAK subfamily (see PAK3; MIM 300142) contain a C-terminal catalytic domain and an N-terminal regulatory domain that has a CDC42 (MIM 116952)-binding domain. In contrast, members of the GCK subfamily (see MAP4K2; MIM 603166), also called the Sps1 subfamily, have an N-terminal catalytic domain and a C-terminal regulatory domain without a CDC42-binding domain. STK24 belongs to the GCK subfamily of STE20-like kinases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MST3
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene STK24
Gene Accession No. B0LT89, Q99KH8, Q9Y6E0
Gene Alias 1810013H02Rik; C76483; epididymis secretory protein Li 95; HEL-S-95; Mammalian STE20-like protein kinase 3; Mammalian STE20-like protein kinase 3 C-terminal; Mammalian STE20-like protein kinase 3 N-terminal; mammalian sterile twenty 3 kinase; Mst3; MST-3; MST3/C; MST3/N; MST3B; OTTHUMP00000018594; RGD1561742; RP11-111L24.5; serine/threonine kinase 24; serine/threonine kinase 24 (STE20 homolog, yeast); serine/threonine-protein kinase 24; Serine/threonine-protein kinase 24 12 kDa subunit; Serine/threonine-protein kinase 24 35 kDa subunit; Serine/threonine-protein kinase 24 36 kDa subunit; STE20; STE20-like kinase 3; STE20-like kinase MST3; sterile 20-like kinase 3; STK24; STK3
Gene Symbols STK24
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues of human Mst3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223255, 361092, 8428
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.