Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MST3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528668
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (96%), Rat (94%), Chicken (85%), Rhesus Monkey (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
STK24 (MST3) is a serine/threonine kinase that is a member of the GCK subfamily of STE20-like proteins. The yeast 'Sterile 20' gene (STE20) functions upstream of the mitogen-activated protein kinase (MAPK) cascade. In mammals, protein kinases related to STE20 can be divided into 2 subfamilies based on their structure and regulation. Members of the PAK subfamily (see PAK3; MIM 300142) contain a C-terminal catalytic domain and an N-terminal regulatory domain that has a CDC42 (MIM 116952)-binding domain. In contrast, members of the GCK subfamily (see MAP4K2; MIM 603166), also called the Sps1 subfamily, have an N-terminal catalytic domain and a C-terminal regulatory domain without a CDC42-binding domain. STK24 belongs to the GCK subfamily of STE20-like kinases.
Specifications
MST3 | |
Polyclonal | |
Unconjugated | |
STK24 | |
1810013H02Rik; C76483; epididymis secretory protein Li 95; HEL-S-95; Mammalian STE20-like protein kinase 3; Mammalian STE20-like protein kinase 3 C-terminal; Mammalian STE20-like protein kinase 3 N-terminal; mammalian sterile twenty 3 kinase; Mst3; MST-3; MST3/C; MST3/N; MST3B; OTTHUMP00000018594; RGD1561742; RP11-111L24.5; serine/threonine kinase 24; serine/threonine kinase 24 (STE20 homolog, yeast); serine/threonine-protein kinase 24; Serine/threonine-protein kinase 24 12 kDa subunit; Serine/threonine-protein kinase 24 35 kDa subunit; Serine/threonine-protein kinase 24 36 kDa subunit; STE20; STE20-like kinase 3; STE20-like kinase MST3; sterile 20-like kinase 3; STK24; STK3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8428 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.97 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y6E0 | |
STK24 | |
Recombinant protein encompassing a sequence within the center region of human MST3. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction