Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MST4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595790 Supplier Invitrogen™ Supplier No. PA595790
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NGAEQDLVQT LSCLSMIITP AFAELKQQDE NNASRNQAIE ELEKSIAVAE AACPGITDKM VKKLIEKFQK CSADESP Positive Samples: Rat thymus; Cellular Location: Cytoplasm, Cytoplasmic side, Golgi apparatus membrane, Peripheral membrane protein.

MST4 is member of the Ste20-like kinase family of serine/threonine kinases and is overexpressed during prostate cancer development. The product of this gene is a member of the GCK group III family of kinases, which are a subset of the Ste20-like kinases. The encoded protein contains an amino-terminal kinase domain, and a carboxy-terminal regulatory domain that mediates homodimerization. The protein kinase localizes to the Golgi apparatus and is specifically activated by binding to the Golgi matrix protein GM130. It is also cleaved by caspase-3 in vitro, and may function in the apoptotic pathway. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MST4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Stk26
Gene Accession No. Q99JT2, Q9P289
Gene Alias 2610018G03Rik; Mammalian STE20-like protein kinase 4; mammalian sterile 20-like 4; Mask; Mst3 and SOK1-related kinase; Mst4; MST-4; OTTHUMP00000215132; OTTHUMP00000215134; RGD1563568; RP6-213H19.1; serine/threonine kinase 26; serine/threonine protein kinase 26; serine/threonine protein kinase MST4; serine/threonine-protein kinase 26; Serine/threonine-protein kinase MASK; serine/threonine-protein kinase MST4; STE20-like kinase 4; STE20-like kinase MST4; STK26
Gene Symbols Stk26
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 340-416 of human STK26 (NP_0576262).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317589, 51765, 70415
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.