Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MST4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111497
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111497 Supplier Invitrogen™ Supplier No. PA5111497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GHSDDESDSE GSDSESTSRE NNTHPEWSFT TVRKKPDPKK VQNGAEQDLV QTLSCLSMII TPAFAELKQQ DENNA Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 92%.

MST4 is member of the Ste20-like kinase family of serine/threonine kinases and is overexpressed during prostate cancer development. The product of this gene is a member of the GCK group III family of kinases, which are a subset of the Ste20-like kinases. The encoded protein contains an amino-terminal kinase domain, and a carboxy-terminal regulatory domain that mediates homodimerization. The protein kinase localizes to the Golgi apparatus and is specifically activated by binding to the Golgi matrix protein GM130. It is also cleaved by caspase-3 in vitro, and may function in the apoptotic pathway. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MST4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Stk26
Gene Accession No. Q9P289
Gene Alias 2610018G03Rik; Mammalian STE20-like protein kinase 4; mammalian sterile 20-like 4; Mask; Mst3 and SOK1-related kinase; Mst4; MST-4; OTTHUMP00000215132; OTTHUMP00000215134; RGD1563568; RP6-213H19.1; serine/threonine kinase 26; serine/threonine protein kinase 26; serine/threonine protein kinase MST4; serine/threonine-protein kinase 26; Serine/threonine-protein kinase MASK; serine/threonine-protein kinase MST4; STE20-like kinase 4; STE20-like kinase MST4; STK26
Gene Symbols Stk26
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human MST4. Recombinant protein control fragment (Product #RP-103363).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51765
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.