Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSTO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552227
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552227 Supplier Invitrogen™ Supplier No. PA552227
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DRACHTSQLT PGTPPPSALH ACTTGEEILA QYLQQQQPGV MSSSHLLLTP CRVAPPYPHL FSSCSPPGMV LDGSPKGAAV ESIPVFGALC SSSSLHQTLE ALARDLTKLD LRRWASFMDA GVEHDDVAEL LQELQSLAQC YQGGDSLVD Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 70%.

MSTO1 is a protein coding gene involved in the regulation of mitochondrial distribution and morphology. Required for mitochondrial fusion and mitochondrial network formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSTO1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MSTO1
Gene Accession No. Q9BUK6
Gene Alias LST005; misato 1, mitochondrial distribution and morphology regulator; misato homolog 1; MST; MSTO1; Protein misato homolog 1; SLTP005
Gene Symbols MSTO1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MSTO1. Recombinant protein control fragment (Product #RP-89152).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55154
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.