Learn More
MSUT2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303993A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
ZC3H14 belongs to the ZC3H14 family.It contains 2 C3H1-type zinc fingers. ZC3H14 binds the polyadenosine RNA oligonucleotides.Specifications
MSUT2 | |
Polyclonal | |
Unconjugated | |
ZC3H14 | |
FLJ11806, mammalian suppressor of tau pathology-2, MSUT-2, nuclear protein UKp68, NY-REN-37, renal carcinoma antigen NY-REN-37, SUT2, UKp68 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79882 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q6PJT7 | |
ZC3H14 | |
Between 686 and 736 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.