Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523176
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523176 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523176 Supplier Invitrogen™ Supplier No. PA523176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The amino acid sequence used as immunogen is 83% homologous in Opossum and Monkey.

MSX1 is a member of the muscle segment homeobox gene family. It functions as a transcriptional repressor during embryogenesis through interactions with components of the core transcription complex and other homeoproteins. MSX1 may also have roles in limb-pattern formation, craniofacial development, particularly odontogenesis, and tumor growth inhibition. Mutations in the gene, which was once known as homeobox 7, have been associated with nonsyndromic cleft lip with or without cleft palate 5, Witkop syndrome, Wolf-Hirschom syndrome, and autosomoal dominant hypodontia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene MSX1
Gene Accession No. P13297, P28360
Gene Alias AA675338; AI324650; ECTD3; homeo box, msh-like 1; homeobox 7; homeobox protein Hox-7; Homeobox protein MSX-1; homeobox, msh-like 1; Hox7; Hox-7; Hox7.1; hox-7.1; HYD1; msh; msh homeo box 1; msh homeobox 1; msh homeobox 1-like protein; msh homeobox homolog 1; Msx1; muscle-segment homeobox; STHAG1
Gene Symbols MSX1
Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human HOX7.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17701, 4487, 81710
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.