Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541436
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541436 Supplier Invitrogen™ Supplier No. PA541436
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MASPSKGNDL FSPDEEGPAV VAGPGPGPGG AEGAAEERRV KVSSLPFSVE Sequence homology: Cow: 100%; Dog: 100%; Goat: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.

Msx2 is a transcriptional repressor whose normal activity may establish a balance between survival and apoptosis of neural crest-derived cells required for proper craniofacial morphogenesis. This protein may also have a role in promoting cell growth under certain conditions and may be an important target for the RAS signaling pathways. Mutations in the Msx2 gene are associated with parietal foramina 1 and craniosynostosis type 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSX2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MSX2
Gene Accession No. P35548
Gene Alias BB122635; CRS2; FPP; homeo box, msh-like 2; homeobox protein Hox-8; Homeobox protein Hox-8-1; homeobox protein MSX-2; homeobox, msh-like 2; HOX8; Hox-8; Hox8.1; Hox-8.1; MSH; msh homeo box 2; msh homeo box homolog 2; msh homeobox 2; msh homeobox homolog 2; MSX2; Msx-2; muscle segmentation homologue (MSX2); PFM; PFM1; Unknown (protein for MGC:142681)
Gene Symbols MSX2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MSX2 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4488
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.