Learn More
Invitrogen™ MSX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578064
Description
Positive Control: HL-60, 293T, A431, HeLa, HepG2, Neuro2A, C8D30, NIH-3T3, Raw264.7, C2C12 Predicted Reactivity: Rat (86%), Dog (86%), Rhesus Monkey (98%), Chimpanzee (100%), Bovine (94%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Msx2 is a transcriptional repressor whose normal activity may establish a balance between survival and apoptosis of neural crest-derived cells required for proper craniofacial morphogenesis. This protein may also have a role in promoting cell growth under certain conditions and may be an important target for the RAS signaling pathways. Mutations in the Msx2 gene are associated with parietal foramina 1 and craniosynostosis type 2.
Specifications
MSX2 | |
Polyclonal | |
Unconjugated | |
MSX2 | |
BB122635; CRS2; FPP; homeo box, msh-like 2; homeobox protein Hox-8; Homeobox protein Hox-8-1; homeobox protein MSX-2; homeobox, msh-like 2; HOX8; Hox-8; Hox8.1; Hox-8.1; MSH; msh homeo box 2; msh homeo box homolog 2; msh homeobox 2; msh homeobox homolog 2; MSX2; Msx-2; muscle segmentation homologue (MSX2); PFM; PFM1; Unknown (protein for MGC:142681) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17702, 4488 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P35548, Q03358 | |
MSX2 | |
Recombinant protein encompassing a sequence within the center region of human MSX2. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.