Learn More
IKA™ MT 40.100 Grinding Chamber
SDP
Catalog No. 01183084 Shop All IKA Works Products
Change view
Click to view available options
Capacity (Metric):
40 mL
Catalog No. Capacity (Metric)
01-183-084 40 mL
1 options
This item is not returnable. View return policy
Catalog No. 01-183-084 Supplier IKA™ Supplier No. 20001173
  • PRODUCT PROMOTION
    $1,654.00
    $1,218.52 / Pack of 100
    Save $435.48
    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
The MT 40.100 disposable grinding chamber with a stainless steel beater reduces soft, medium, hard and brittle materials with a Mohs hardness of up to 5 (manganese or apatite: 5 Mohs).
The chamber is made of transparent plastic so grinding tests can be observed at any time
Specifications
Product Type Grinding Chamber
Capacity (Metric) 40 mL