Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MT-ATP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575605
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575605 Supplier Invitrogen™ Supplier No. PA575605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Mitochondrial ATP synthase is composed of two multi-subunit complexes that utilize an inner membrane electrochemical gradient to catalyze the synthesis of ATP during oxidative phosphorylation. The two multi-subunit complexes are designated F1 and F0, the former of which comprises the soluble catalytic core and the latter of which comprises the membrane-spanning proton channel of ATP synthase. ATP8, also known as MT-ATP8, ATPASE8 or MTATP8, is a 68 amino acid single-pass membrane protein that localizes to mitochondria and functions as a minor subunit of the F0 complex. The gene encoding ATP8 maps to the mitochondrial chromosome and may be involved in the pathogenesis of multiple sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT-ATP8
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MT-ATP8
Gene Accession No. P03928, P03930, P11608
Gene Alias A6L; ATP synthase F0 subunit 8; ATP synthase protein 8; ATP8; Atpase8; Chargerin II; F-ATPase subunit 8; mitochondrially encoded ATP synthase 8; Mtatp8; MT-ATP8
Gene Symbols MT-ATP8
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human MT-ATP8.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17706, 26196, 4509
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.