Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MT-CO2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501570119
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0119 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0119 Supplier Bethyl Laboratories, Inc Supplier No. A305318A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla and Chimpanzee

Cytochrome c oxidase is the fourth complex in the respiratory chain and is responsible for catalyzing the conversion of O2 to H2O. Subunit 2 of the cytochrome c oxidase complex combines with two other subunits (1 and 3) to form a core protein structure that performs many functions of the enzyme. The metallic copper center of this subunit transfers electrons to the heme center of subunit 1, which results in the movement of electrons from cytochrome c to the heme A3 and copper B metallic center of complex IV. Defects in subunit 2 of cytochrome c oxidase can result in COX deficiency, which causes a wide range of symptoms from local myopathy to multiple system pathologies that begin between infancy and adulthood. Abnormalities in this subunit are also associated with tumor development.

Specifications

Antigen MT-CO2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene MT-CO2
Gene Accession No. P00403
Gene Alias MTCO2, MT-CO2, Cytochrome c oxidase 2, Cytochrome c oxidase II, Cytochrome c oxidase polypeptide II, COII, COXII
Gene Symbols MT-CO2
Host Species Rabbit
Immunogen Between 125 and 175
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4513
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.