Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MT-CYB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100740 Supplier Invitrogen™ Supplier No. PA5100740
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MT-CYB.

CYTB belongs to the cytochrome b family. It is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is a respiratory chain that generates an electrochemical potential coupled to ATP synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT-CYB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MT-CYB
Gene Accession No. P00156, P00158, P00159
Gene Alias COB; Complex III subunit 3; Complex III subunit III; CYTB; Cytochrome b; Cytochrome b-c1 complex subunit 3; mitochondrially encoded cytochrome b; Mtcyb; Mt-Cyb; mt-Cytb; Ubiquinol-cytochrome-c reductase complex cytochrome b subunit; UQCR3
Gene Symbols CYTB, MT-CYB
Host Species Rabbit
Immunogen A synthesized peptide derived from human MT-CYB(Accession P00156), corresponding to amino acid residues M1-T47.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17711, 26192, 4519
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.