Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MT-ND1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120599
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120599 Supplier Invitrogen™ Supplier No. PA5120599
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, HepG2, 293T, mouse heart, mouse brain, mouse kidney, rat heart, rat brain. Immunogen sequence: MPMANLLLLI VPILIAMAFL MLTERKILGY MQLRKGPNVV GPYGLLQPFA DAMKLFTKEP LKPATSTITL YITAPTLALT IALLLWTPLP MPNPLVNLNL.

A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 1 (ND1) binds rotenone and rotenone analogs and might be involved in electron transfer to ubiquinone. Mutations in the ND1 gene may be implicated in several disorders, including Leber hereditary optic neuropathy, Alzheimer's disease and Parkinson disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT-ND1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene mt-Nd1
Gene Accession No. P03886, P03888, P03889
Gene Alias mitochondrially encoded NADH dehydrogenase 1; Mtnd1; MT-ND1; NADH dehydrogenase; NADH dehydrogenase subunit 1; NADH1; NADH-ubiquinone oxidoreductase chain 1; Nd1
Gene Symbols mt-Nd1
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17716, 26193, 4535
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.