Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MT-ND2, Mouse anti-Human, Clone: 9E12-1B3, Millipore Sigma™

Catalog No. MABS204725 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS204725 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS204725 Supplier MilliporeSigma Supplier No. MABS204725UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MT-ND2, clone 9E12-1B3, Cat. No. MABS2047, is a mouse monoclonal antibody that detects NADH-ubiquinone oxidoreductase chain 2 and has been tested for use in Western Blotting.

NADH-ubiquinone oxidoreductase chain 2 (UniProt: P03891; also known as EC: 1.6.5.3, NADH dehydrogenase subunit 2, MT-ND2) is encoded by the MT-ND2 (also known as MTND2, NADH2, ND2) gene (Gene ID: 4536) in human. MT-ND2 is a mitochondrial inner membrane protein that forms the core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I). It is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone (Coenzyme Q10) that participates in the generation of a proton gradient, which is then used for ATP synthesis. Defects in MT-ND2 gene are known to cause of Leber hereditary optic neuropathy (LHON) that is a maternally inherited disease resulting in acute or subacute loss of central vision, due to optic nerve dysfunction. LHON results from primary mitochondrial DNA mutations affecting the respiratory chain complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT-ND2
Applications Western Blot
Classification Monoclonal
Clone 9E12-1B3
Conjugate Unconjugated
Dilution Western Blotting Analysis: 2μg/mL from a representative lot detected MT-ND2 in mitochondria from human neonatal dermal fibroblasts and mitochondria from human neonatal dermal fibroblasts depleted of mtDNA (Courtesy of Michael F. Marusich, Ph.D., mAbDx, Inc., Eugene, OR USA).
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing HEPES-Buffered Saline (150 mM NaCl, 15 mM HEPES, pH 7.2) with 0.02% sodium azide.
Gene Alias NADH-ubiquinone oxidoreductase chain 2;EC: 1.6.5.3;NADH dehydrogenase subunit 2;MT-ND2
Gene Symbols MT-ND2;MTND2;NADH2;ND2
Host Species Mouse
Immunogen Synthetic peptide corresponding to the N-terminus of human MT-ND2.
Purification Method Protein L
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 9E12-1B3 detects NADH-ubiquinone oxidoreductase chain 2 (MT-ND2) in mitochondria from human cells. It targets an epitope within the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.