Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MT-ND3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | MT-ND3 |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
MT-ND3 Polyclonal specifically detects MT-ND3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MT-ND3 | |
Polyclonal | |
Rabbit | |
Human | |
P03897 | |
4537 | |
This antibody was developed against a recombinant protein corresponding to amino acids: GYIEKSTPYECGFDPISPARVPFSIK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Complex I ND3 Subunit, EC 1.6.5.3, Mitochondrially Encoded NADH Dehydrogenase 3, MTND3, NADH Dehydrogenase 3, NADH Dehydrogenase Subunit 3, NADH Dehydrogenase, Subunit 3 (Complex I), NADH3, NADH-Ubiquinone Oxidoreductase Chain 3, ND3 | |
ND3 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title