Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MT-ND4, Mouse anti-Human, Clone: 9E4-2D8, Millipore Sigma™

Catalog No. MABS199425 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS199425 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS199425 Supplier MilliporeSigma Supplier No. MABS199425UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MT-ND4, clone 9E4-2D8, Cat. No. MABS1994,is a mouse monoclonal antibody that detects NADH-ubiquinone oxidoreductase chain 4 and has been tested for use in Western Blotting.

NADH-ubiquinone oxidoreductase chain 4 (UniProt: P03905; also known as EC: 1.6.5.3, NADH dehydrogenase subunit 4, MT-ND4) is encoded by the MT-ND4 (also known as MTND4, NADH4, ND4) gene (Gene ID: 4538) in human. MT-ND4 is an inner mitochondrial membrane protein that belongs to the complex I subunit 4 family. It is a core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I). The complex couples the oxidation of NADH and the reduction of ubiquinone (Coenzyme Q10), to the generation of a proton gradient, which is then used for ATP synthesis. The complex can be dissociated into two main sub-complexes, corresponding to the "ankle" of the boot, and the "foot" of the boot. The ankle is thought to protrude from the membrane so as to be predominantly in the aqueous phase on the matrix side. It contains the binding site for NAD(H), and the input electron transfer chain. The foot (hydrophobic) is membrane bound and contains a catalytic site at which ubiquinone is reduced. Defects in MT-ND4 gene are known to cause Leber hereditary optic neuropathy (LHON) that is characterized by acute or subacute loss of central vision due to optic nerve dysfunction. LHON results from primary mitochondrial DNA mutations affecting the respiratory chain complexes. Other diseases associated with MT-ND4 gene mutations are age-related macular degeneration, mesial temporal lobe epilepsy, and cystic fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT-ND4
Applications Western Blot
Classification Monoclonal
Clone 9E4-2D8
Conjugate Unconjugated
Dilution Western Blotting Analysis: 4μg/mL from a representative lot detected MT-ND4 in mitochondria from Human Neonatal Dermal Fibroblasts and mitochondria from Human neonatal dermal fibroblasts depleted of mtDNA. (Courtesy of  Michael F. Marusich, Ph.D., mAbDx, Inc., Eugene, OR, USA ).
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing HEPES-Buffered Saline (150 mM NaCl, 15 mM HEPES, pH 7.2) with 0.02% sodium azide.
Gene Alias NADH-ubiquinone oxidoreductase chain 4;EC: 1.6.5.3;NADH dehydrogenase subunit 4
Gene Symbols MT-ND4;MTND4;NADH4;ND4
Host Species Mouse
Immunogen KLH-conjugated linear peptide from the C-terminal region.
Purification Method Protein L
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 9E4-2D8 detects NADH-ubiquinone oxidoreductase chain 4 in mitochodria isolated from human cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.