Learn More
Invitrogen™ MT-ND4 Polyclonal Antibody

Description
Positive Samples: NIH/3T3, PC-12, 293T, Mouse heart, Mouse kidney, Mouse liver, Mouse brain, Rat heart; Cellular Location: Mitochondrion membrane, Multi-pass membrane protein Immunogen sequence: TTQRGKLTNH MINLQPS.
Specifications
Specifications
| Antigen | MT-ND4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.41 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | MT-ND4 |
| Gene Accession No. | P03905, P03911, P05508 |
| Gene Alias | mitochondrially encoded NADH dehydrogenase 4; Mtnd4; MT-ND4; NADH dehydrogenase subunit 4; NADH4; NADH-ubiquinone oxidoreductase chain 4; ND4 |
| Gene Symbols | MT-ND4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.