Learn More
Invitrogen™ MT-ND4 Polyclonal Antibody

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human HepG2 whole cell, human THP-1 whole cell, human Caco-2 whole cell, human U2OS whole cell, human A549 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | MT-ND4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and 0.05mg sodium azide |
| Gene | MT-ND4 |
| Gene Accession No. | P03905 |
| Gene Alias | mitochondrially encoded NADH dehydrogenase 4; Mtnd4; MT-ND4; NADH dehydrogenase subunit 4; NADH4; NADH-ubiquinone oxidoreductase chain 4; ND4 |
| Gene Symbols | MT-ND4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.