Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MT-ND4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116791
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116791 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116791 Supplier Invitrogen™ Supplier No. PA5116791
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SGC-7901, U-87MG Immunogen sequence: LVTALYSLYM FTTTQWGSLT HHINNMKPSF TRENTLMFMH LSPILLLSLN PDIITGFSS.

Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT-ND4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene MT-ND4
Gene Accession No. P03905, P03911, P05508
Gene Alias mitochondrially encoded NADH dehydrogenase 4; Mtnd4; MT-ND4; NADH dehydrogenase subunit 4; NADH4; NADH-ubiquinone oxidoreductase chain 4; ND4
Gene Symbols MT-ND4
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17719, 26201, 4538
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.