Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MT-ND4 Polyclonal Antibody
MT-ND4 Polyclonal Antibody
Supplier: Thermo Scientific PA576458
Description
MT-ND4 Polyclonal Antibody for Western Blot

Specifications
MT-ND4 | |
Polyclonal | |
Unconjugated | |
MT-ND4 | |
mitochondrially encoded NADH dehydrogenase 4; Mtnd4; MT-ND4; NADH dehydrogenase subunit 4; NADH4; NADH-ubiquinone oxidoreductase chain 4; ND4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17719, 26201, 4538 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P03905, P03911, P05508 | |
MT-ND4 | |
Synthetic peptide of mouse MT-ND4. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction