Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MT-ND5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572501
Description
MT-ND5 Polyclonal Antibody for Western Blot, IHC (P), Flow
A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 5 (ND5) is most likely a component of the hydrophobic protein fragment of Complex I. Mutations in the gene encoding for ND5 are implicated in Mitochondrial neurogastrointestinal encephalomyopathy, an autosomal recessive multisystem disorder that results in dramatically elevated levels of circulating thymidine and deoxyuridine.
Specifications
MT-ND5 | |
Polyclonal | |
Unconjugated | |
MT-ND5 | |
mitochondrially encoded NADH dehydrogenase 5; MTND5; mt-Nd5; NADH Dehydrogenase 5; NADH dehydrogenase subunit 5; NADH5; NADH-ubiquinone oxidoreductase chain 5; ND5 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
4540 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P03915 | |
MT-ND5 | |
KLH conjugated synthetic peptide between 544-570 amino acids from the C-terminal region of human ND5. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction