Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MT-ND6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103954
Description
Antibody detects endogenous levels of total MT-ND6.
Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
MT-ND6 | |
Polyclonal | |
Unconjugated | |
mt-Nd6 | |
mitochondrially encoded NADH dehydrogenase 6; MTND6; MT-ND6; NADH dehydrogenase subunit 6; NADH6; NADH-ubiquinone oxidoreductase chain 6; ND6 | |
Rabbit | |
Affinity chromatography | |
RUO | |
17722, 26203, 4541 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P03923, P03925, P03926 | |
mt-Nd6, ND6 | |
A synthesized peptide derived from human MT-ND6(Accession P03923), corresponding to amino acid residues M54-L104. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction