Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MT1E Monoclonal Antibody (C1), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543655
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543655 100 μL
PIMA543656 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543655 Supplier Invitrogen™ Supplier No. MA543655
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MT1E is a protein coding gene. Metallothioneins have a high content of cysteine residues that bind various heavy metals; these proteins are transcriptionally regulated by both heavy metals and glucocorticoids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT1E
Applications Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene MT1E
Gene Accession No. P04732
Gene Alias metallothionein 1E; metallothionein 1E (functional); metallothionein D; metallothionein-1E; Metallothionein-IE; MT1; MT1E; MT-1E; MTD; MT-IE
Gene Symbols MT1E
Host Species Mouse
Immunogen Recombinant MT1E (Met1-Ala61) expressed in E.coli.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4493
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.