Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTA2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$322.50 - $534.50
Specifications
Antigen | MTA2 |
---|---|
Dilution | Western Blot 0.04-0.4 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MTA2 Polyclonal antibody specifically detects MTA2 in Human samples. It is validated for Western BlotSpecifications
MTA2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Chromatin Research, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
PBS, pH 7.2, 40% glycerol | |
9219 | |
IgG | |
Affinity purified |
Western Blot 0.04-0.4 ug/mL | |
Polyclonal | |
Purified | |
RUO | |
Human | |
DKFZp686F2281, metastasis associated 1 family, member 2, metastasis -associated gene 1-like 1, metastasis associated gene family, member 2, Metastasis-associated 1-like 1, metastasis-associated protein 2, metastasis-associated protein MTA2, MTA1-L1 protein, MTA1L1MTA1-L1, p53 target protein in deacetylase complex, PID | |
This antibody was developed against Recombinant Protein corresponding to amino acids: KTPTQLEGATRGTTEPHSRGHLSRPEAQSLSPYTTSANRAKLLAKNRQTFLLQTTK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title