Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTA2 Monoclonal Antibody (17A1)
GREENER_CHOICE

Catalog No. PIMA534950
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534950 Supplier Invitrogen™ Supplier No. MA534950
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: K562, MCF-7, SH-SY5Y, Daudi, human colon carcinoma tissue, human skin tissue, human breast tissue, human breast carcinoma tissue, human gastric carcinoma tissue, human small intestine tissue.

This gene encodes a protein that has been identified as a component of NuRD, a nucleosome remodeling deacetylase complex identified in the nucleus of human cells. It shows a very broad expression pattern and is strongly expressed in many tissues. It may represent one member of a small gene family that encode different but related proteins involved either directly or indirectly in transcriptional regulation. Their indirect effects on transcriptional regulation may include chromatin remodeling. It is closely related to another member of this family, a protein that has been correlated with the metastatic potential of certain carcinomas. These two proteins are so closely related that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. One of the proteins known to be a target protein for this gene product is p53. Deacetylation of p53 is correlated with a loss of growth inhibition in transformed cells supporting a connection between these gene family members and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTA2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 17A1
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene Mta2
Gene Accession No. O94776
Gene Alias AW550797; DKFZp686F2281; Mata1l1; metastais-associated gene family, member 2; metastasis associated 1 family member 2; metastasis associated 1 family, member 2; metastasis associated 1-like 1; metastasis -associated gene 1-like 1; metastasis associated gene family, member 2; metastasis-associated 1-like 1; metastasis-associated gene family, member 2; metastasis-associated protein 2; metastasis-associated protein MTA2; mmta2; Mta1l1; MTA1-L1 protein; Mta2; p53 target protein in deacetylase complex; PID
Gene Symbols Mta2
Host Species Mouse
Immunogen Recombinant protein within Human MTA2 aa 77-286.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9219
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.