Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTA3 Monoclonal Antibody (OTI2B8)
GREENER_CHOICE

Catalog No. PIMA527203
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527203 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527203 Supplier Invitrogen™ Supplier No. MA527203
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MTA3 Monoclonal Antibody for Western Blot

Plays a role in maintenance of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels. Contributes to transcriptional repression by BCL6. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTA3
Applications Western Blot
Classification Monoclonal
Clone OTI2B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Mta3
Gene Accession No. Q9BTC8
Gene Alias 1110002J22Rik; KIAA1266; metastasis associated 1 family member 3; metastasis associated 1 family, member 3; metastasis associated 3; metastasis associated family, member 3; metastasis associated gene family, member 3; metastasis-associated protein MTA3; mKIAA1266; Mta3
Gene Symbols Mta3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-259 of MTA3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57504
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.