Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTAP Mouse anti-Human, Mouse, Rat, Clone: 2B1G6, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | MTAP |
---|---|
Clone | 2B1G6 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7121
![]() ![]() |
Proteintech Group Inc
667061IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes an enzyme that plays a major role in polyamine metabolism and is important for the salvage of both adenine and methionine. The encoded enzyme is deficient in many cancers because this gene and the tumor suppressor p16 gene are co-deleted. Multiple alternatively spliced transcript variants have been described for this gene, but their full-length natures remain unknown.Specifications
MTAP | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
c86fus, MSAP, MTA phosphorylase, MTAP, MTAPase | |
MTAP | |
IgG1 | |
Protein G | |
Antibody |
2B1G6 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q13126, Q9CQ65 | |
298227, 4507, 66902 | |
MTAP Fusion Protein Ag17954 | |
Primary | |
-20°C | |
MTAP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title