Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTAP Mouse anti-Human, Mouse, Rat, HRP, Clone: 2B1G6, Proteintech
SDP

Catalog No. 501737243 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7243 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7243 Supplier Proteintech Group Inc Supplier No. HRP66706100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes an enzyme that plays a major role in polyamine metabolism and is important for the salvage of both adenine and methionine. The encoded enzyme is deficient in many cancers because this gene and the tumor suppressor p16 gene are co-deleted. Multiple alternatively spliced transcript variants have been described for this gene, but their full-length natures remain unknown.

Specifications

Antigen MTAP
Applications Western Blot
Classification Monoclonal
Clone 2B1G6
Concentration 0.67 mg/mL
Conjugate HRP
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MTAP
Gene Accession No. Q13126, Q9CQ65
Gene Alias c86fus, MSAP, MTA phosphorylase, MTAP, MTAPase
Gene Symbols MTAP
Host Species Mouse
Immunogen MTAP Fusion Protein Ag17954
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298227, 4507, 66902
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.