Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTAP Mouse anti-Human, Mouse, Rat, Unconjugated, Clone: 2G4, Abnova™

Catalog No. 89335056 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-056 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-056 Supplier Abnova Corporation Supplier No. H00004507M01J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

MTAP Monoclonal antibody specifically detects MTAP in Human, Mouse, Rat samples. It is validated for WB-Ce, IF, S-ELISA, ELISA, WB-Re.

Specifications

Antigen MTAP
Applications Immunofluorescence, ELISA, Western Blot
Classification Monoclonal
Clone 2G4
Conjugate Unconjugated
Description methylthioadenosine phosphorylase
Gene MTAP
Gene Accession No. BC018625
Gene Alias MSAP, c86fus
Host Species Mouse
Immunogen MTAP (AAH18625, 1 a.a. ∼ 154 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4507
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.