Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTAP Mouse anti-Human, Unconjugated, Clone: MTAP/1813, Abnova™

Catalog No. 89337803 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-803 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-803 Supplier Abnova Corporation Supplier No. MAB21312
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

MTAP Monoclonal antibody specifically detects MTAP in Human samples. It is validated for ELISA, WB-Ce, IHC-P.

Specifications

Antigen MTAP
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MTAP/1813
Conjugate Unconjugated
Description methylthioadenosine phosphorylase
Gene MTAP
Gene Alias MSAP, c86fus
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 97-196 of human MTAP
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4507
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.