Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTAP Rabbit anti-Human, Clone: MTAP/3137R, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$278.72

Specifications

Antigen MTAP
Clone MTAP/3137R
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121914
SDP
View Documents
Novus Biologicals
NBP30722020UG
20 μg
Each of 1 for $278.72
Only null left
Add to Cart
 
Description

Description

MTAP Monoclonal specifically detects MTAP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.
Specifications

Specifications

MTAP
0.2 mg/ml
Western Blot, ELISA, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
5'-methylthioadenosine phosphorylase, c86fus, EC 2.4.2.28, methylthioadenosine phosphorylase, MSAPMeSAdo phosphorylase, MTA phosphorylase, MTAPase, S-methyl-5'-thioadenosine phosphorylase
Recombinant human MTAP protein fragment (aa97-196) (exact sequence is proprietary)
Primary
Store at 4°C.
MTAP/3137R
Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
4507
IgG
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.